Tetrakis(2-ethylhexyl) Orthosilicate ≥97.0% (by GC)

Supplier: TCI America
Warning

T3748-25ML T3748-5ML
TCT3748-25MLEA 116 USD
TCT3748-25ML TCT3748-5ML
Tetrakis(2-ethylhexyl) Orthosilicate ≥97.0% (by GC)
Tetrakis(2-ethylhexyl) Orthosilicate
Formula: C₃₂H₆₈O₄Si
Boiling Pt: 365 °C
Flash Pt: 199 °C
MDL Number: MFCD00015263
CAS Number: 115-82-2
EINECS: 204-109-6

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR