3-Hydroxy-1-methacryloyloxyadamantane ≥99.0% (by GC), purified by sublimation

Supplier: TCI America
Warning

Synonyms: 3-Hydroxy-1-adamantyl methacrylate, 3-Hydroxyadamantan-1-yl methacrylate, Methacrylic acid 3-Hydroxy-1-adamantyl ester

H1689-1G H1689-5G
TCH1689-1GEA 104 USD
TCH1689-1G TCH1689-5G
3-Hydroxy-1-methacryloyloxyadamantane ≥99.0% (by GC), purified by sublimation
3-Hydroxy-1-methacryloyloxyadamantane
Formula: C₁₄H₂₀O₃
MW: 236.31 g/mol
MDL Number: MFCD13152044
CAS Number: 115372-36-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR