6,6'-Dibromo-1,1'-bis(2-exylhexyl)isoindigo ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D5766-1G D5766-200MG
TCD5766-1GEA 339 USD
TCD5766-1G TCD5766-200MG
6,6'-Dibromo-1,1'-bis(2-exylhexyl)isoindigo ≥98.0% (by HPLC, total nitrogen)
6,6'-Dibromo-1,1'-bis(2-exylhexyl)isoindigo
Formula: C₃₂H₄₀Br₂N₂O₂
MDL Number: MFCD28125575
CAS Number: 1147124-23-9

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR