2-(2,6-Diisopropylphenyl)-3,3-dimethyl-2-azaspiro[4.5]dec-1-en-2-ium Hydrogen Dichloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Cyclohexyl-CAAC

D5596-1G D5596-5G
TCD5596-1GEA 149 USD
TCD5596-1G TCD5596-5G
2-(2,6-Diisopropylphenyl)-3,3-dimethyl-2-azaspiro[4.5]dec-1-en-2-ium Hydrogen Dichloride ≥98.0% (by HPLC, titration analysis)
2-(2,6-Diisopropylphenyl)-3,3-dimethyl-2-azaspiro[4.5]dec-1-en-2-ium Hydrogen Dichloride
Formula: C₂₃H₃₇Cl₂N
MW: 398.46 g/mol
MDL Number: MFCD20922893
CAS Number: 1141464-90-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR