Estradiol Dipropionate ≥98.0% (by GC)

Supplier: TCI America
Warning

E1344-1G E1344-250MG
TCE1344-1GEA 239 USD
TCE1344-1G TCE1344-250MG
Estradiol Dipropionate ≥98.0% (by GC)
Estradiol Dipropionate
Formula: C₂₄H₃₂O₄
MDL Number: MFCD00051146
CAS Number: 113-38-2
EINECS: 204-026-5
Merck Index: 14,03703

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C
Specific rotation [a]20/D +38.0 to +42.0 deg (C=1, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR