Dimethylphenylvinylsilane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Dimethyl(phenyl)(vinyl)silane, Vinylphenyldimethylsilane

D5645-1G D5645-5G
TCD5645-1GEA 19 USD
TCD5645-1G TCD5645-5G
Dimethylphenylvinylsilane ≥98.0% (by GC)
Dimethylphenylvinylsilane
Formula: C₁₀H₁₄Si
MW: 162.31 g/mol
Boiling Pt: 82 °C
Flash Pt: 59 °C
MDL Number: MFCD00053857
CAS Number: 1125-26-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR