Potassium trifluoro(2-fluoropyridin-3-yl)borate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Potassium 2-Fluoropyridine-3-trifluoroborate

P2675-1G P2675-5G
TCP2675-1GEA 66 USD
TCP2675-1G TCP2675-5G
Potassium trifluoro(2-fluoropyridin-3-yl)borate ≥98.0% (by HPLC, titration analysis)
Potassium trifluoro(2-fluoropyridin-3-yl)borate
Formula: C₅H₃BF₄NK
MW: 202.99 g/mol
MDL Number: MFCD09993002
CAS Number: 1111732-91-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR