2-Methyl-4,6-bis[(n-octylthio)methyl]phenol ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-Methyl-4,6-bis(n-octylsulphanylmethyl)phenol, 2-Methyl-4,6-bis((octylthio)methyl)phenol

M3227-25G M3227-500G
TCM3227-25GEA 34 USD
TCM3227-25G TCM3227-500G
2-Methyl-4,6-bis[(n-octylthio)methyl]phenol ≥97.0% (by GC)
2-Methyl-4,6-bis[(n-octylthio)methyl]phenol
Formula: C₂₅H₄₄OS₂
MW: 424.75 g/mol
MDL Number: MFCD00134699
CAS Number: 110553-27-0
EINECS: 402-860-6

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR