Pyritinol ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5,5'-(Disulphanediylbis(methylene))bis(4-(hydroxymethyl)-2-methylpyridin-3-ol), Pyrithioxin, Pyridoxine Disulphide

P2250-25G P2250-5G
TCP2250-25GEA 571 USD
TCP2250-25G TCP2250-5G
Pyritinol ≥96.0% (by HPLC, titration analysis)
Pyritinol
Formula: C₁₆H₂₀N₂O₄S₂
MW: 368.47 g/mol
MDL Number: MFCD00151477
CAS Number: 1098-97-1
EINECS: 214-150-1
Merck Index: 14,07996

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR