2,4,4-Trimethylpentan-2-amine hydrobromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: tert-Octylammonium bromide, 1,1,3,3-Tetramethylbutylamine hydrobromide, tert-Octylamine hydrobromide

T3783-1G T3783-5G
TCT3783-1GEA 34 USD
TCT3783-1G TCT3783-5G
2,4,4-Trimethylpentan-2-amine hydrobromide ≥97.0% (by titrimetric analysis)
2,4,4-Trimethylpentan-2-amine hydrobromide
Formula: C₈H₁₉N·HBr
MW: 210.16 g/mol
CAS Number: 1093859-61-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR