Formaldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
F0323-10G
F0323-1G
TCF0323-10GEA
178
USD
TCF0323-10G
TCF0323-1G
Formaldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)
Formaldehyde 2,4-Dinitrophenylhydrazone
Formula:
C₇H₆N₄O₄ MW: 210.15 g/mol |
MDL Number:
MFCD00191364 CAS Number: 1081-15-8 UN: 1325 ADR: 4.1,III |
Specification Test Results
Appearance | Orange to Brown powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 165.0 to 168.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...