Tetradecamethylhexasiloxane ≥96.0% (by GC)

Supplier: TCI America

T3685-25ML T3685-5ML
TCT3685-25MLEA 222 USD
TCT3685-25ML TCT3685-5ML
Tetradecamethylhexasiloxane ≥96.0% (by GC)
Tetradecamethylhexasiloxane
Formula: C₁₄H₄₂O₅Si₆
Boiling Pt: 142 °C
Melting Pt: −59 °C
Flash Pt: 118 °C
MDL Number: MFCD08275354
CAS Number: 107-52-8
EINECS: 203-499-5
Merck Index: 14,09197

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR