N-Methoxy-N-methylpropionamide ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: N-Methoxy-N-methylpropanamide

M3096-1G M3096-5G
TCM3096-1GEA 55 USD
TCM3096-1G TCM3096-5G
N-Methoxy-N-methylpropionamide ≥97.0% (by GC)
N-Methoxy-N-methylpropionamide
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD09833541
CAS Number: 104863-65-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR