3-Iodobenzamide ≥98.0% (by GC)

Supplier: TCI America
Warning

I1035-1G I1035-5G
TCI1035-1GEA 44 USD
TCI1035-1G TCI1035-5G
3-Iodobenzamide ≥98.0% (by GC)
3-Iodobenzamide
Formula: C₇H₆INO
MW: 247.04 g/mol
MDL Number: MFCD00143169
CAS Number: 10388-19-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 185.0 to 189.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR