Isopentylamine hydroiodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3-Methylbutan-1-amine Hydroiodide, Isopentylammonium iodide

I1095-1G I1095-5G
TCI1095-1GEA 36 USD
TCI1095-1G TCI1095-5G
Isopentylamine hydroiodide ≥98.0% (by titrimetric analysis)
Isopentylamine hydroiodide
Formula: C₅H₁₃N·HI
MW: 215.08 g/mol
MDL Number: MFCD33021473
CAS Number: 2733412-76-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR