Isopentylamine hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

I1094-1G I1094-5G
TCI1094-1GEA 41 USD
TCI1094-1G TCI1094-5G
Isopentylamine hydrobromide ≥98.0% (by titrimetric analysis)
Isopentylamine hydrobromide
Formula: C₅H₁₃N·HBr
MDL Number: MFCD33021450
CAS Number: 2733412-57-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR