Formamidinium tetrafluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Danger

Synonyms: Formamidine tetrafluoroborate

F1152-1G F1152-5G
TCF1152-1GEA 61 USD
TCF1152-1G TCF1152-5G
Formamidinium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Formamidinium tetrafluoroborate
Formula: CH₅BF₄N₂
MW: 131.87 g/mol
CAS Number: 2607106-18-1
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR