N,N-Dimethyl-1,3-propanediamine dihydroiodide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: N,N-Dimethyl-1,3-propanediammonium Diiodide, N1,N1-Dimethylpropane-1,3-diamine dihydroiodide, 3-(Dimethylamino)propylamine dihydroiodide

D5619-1G D5619-5G
TCD5619-1GEA 42 USD
TCD5619-1G TCD5619-5G
N,N-Dimethyl-1,3-propanediamine dihydroiodide ≥97.0% (by titrimetric analysis)
N,N-Dimethyl-1,3-propanediamine dihydroiodide
Formula: C₅H₁₄N₂·₂HI
MW: 358.01 g/mol
CAS Number: 2561497-43-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR