N,N-Dimethyl-1,3-propanediamine dihydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: N1,N1-Dimethylpropane-1,3-diamine dihydrobromide, N,N-Dimethyl-1,3-propanediammonium Dibromide, 3-(Dimethylamino)propylamine dihydrobromide

D5618-1G D5618-5G
TCD5618-1GEA 34 USD
TCD5618-1G TCD5618-5G
N,N-Dimethyl-1,3-propanediamine dihydrobromide ≥98.0% (by titrimetric analysis)
N,N-Dimethyl-1,3-propanediamine dihydrobromide
Formula: C₅H₁₄N₂·₂HBr
MW: 264.01 g/mol
CAS Number: 2710685-13-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR