Fenspiride hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 8-Phenethyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one hydrochloride, Fenspiride HCl

F1211-1G F1211-5G
TCF1211-1GEA 59 USD
TCF1211-1G TCF1211-5G
Fenspiride hydrochloride ≥98.0% (by HPLC, titration analysis)
Fenspiride hydrochloride
Formula: C₁₅H₂₀N₂O₂·HCl
MW: 296.80 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133315
CAS Number: 5053-08-7
EINECS: 225-752-9
Merck Index: 14,03999

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR