N-(1,1,3,3-Tetramethylbutyl)acrylamide ≥95.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

T3861-100G T3861-25G
TCT3861-100GEA 101 USD
TCT3861-100G TCT3861-25G
N-(1,1,3,3-Tetramethylbutyl)acrylamide ≥95.0% (by GC) stabilized
N-(1,1,3,3-Tetramethylbutyl)acrylamide
Formula: C₁₁H₂₁NO
Boiling Pt: 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048259
CAS Number: 4223-03-4
EINECS: 224-169-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR