2-Phenylethyl methacrylate ≥98.0% (by GC) stabilised

Supplier: TCI America

Synonyms: Phenethyl methacrylate, Methacrylic acid 2-Phenylethyl ester, Methacrylic acid Phenethyl ester

Stabilization: Stabilised with hydroquinone (HQ)

P2714-25G P2714-5G
TCP2714-25GEA 195 USD
TCP2714-25G TCP2714-5G
2-Phenylethyl methacrylate ≥98.0% (by GC) stabilised
2-Phenylethyl methacrylate
Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Boiling Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038232
CAS Number: 3683-12-3
EINECS: 222-969-0

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR