4-Fluoroaniline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

F1271-25G F1271-5G
TCF1271-25GEA 286 USD
TCF1271-25G TCF1271-5G
4-Fluoroaniline hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluoroaniline hydrochloride
Formula: C₆H₆FN·HCl
Boiling Pt: 167 °C
Melting Pt: 248 °C
MDL Number: MFCD00035500
CAS Number: 2146-07-8

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR