IC261 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3-(2,4,6-Ttrimethoxybenzylidene)indolin-2-one, (E)-3-(2,4,6-trimethoxybenzylidene)indolin-2-one, SU-5607

I1077-10MG I1077-50MG
TCI1077-10MGEA 120 USD
TCI1077-10MG TCI1077-50MG
IC261 ≥98.0% (by HPLC)
IC261
Formula: C₁₈H₁₇NO₄
MW: 311.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00118156
CAS Number: 186611-52-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 212.0 to 216.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR