K02288 ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 3-[6-Amino-5-(3,4,5-trimethoxyphenyl)pyridin-3-yl]phenol

K0065-10MG K0065-50MG
TCK0065-10MGEA 137 USD
TCK0065-10MG TCK0065-50MG
K02288 ≥98.0% (by HPLC)
K02288
Formula: C₂₀H₂₀N₂O₄
MW: 352.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26936347
CAS Number: 1431985-92-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.50 to 8.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR