Plerixafor (AMD3100) ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
AMD 3100, Mezobil, JM 3100, Plerixafor, 1,4-Bis((1,4,8,11-tetraazacyclotetradecan-1-yl)methyl)benzene, 1,1'-(1,4-phenylenebis(methylene))bis(1,4,8,11-tetraazacyclotetradecane)octahydrochloride dihydrate, 1,4-Bis[(1,4,8,11-tetraazacyclotetradecan-1-yl)methyl]benzene
P2678-250MG
P2678-50MG
TCP2678-250MGEA
298
USD
TCP2678-250MG
TCP2678-50MG
Plerixafor (AMD3100) ≥98.0% (by GC)
Plerixafor (AMD3100)
Formula:
C₂₈H₅₄N₈ MW: 502.78 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05662218 CAS Number: 110078-46-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Water | max. 13.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...