Quinoline-N-oxide ≥98.0% (by GC)

Supplier: TCI America

Q0104-25G Q0104-5G
TCQ0104-25GEA 213 USD
TCQ0104-25G TCQ0104-5G
Quinoline-N-oxide ≥98.0% (by GC)
Quinoline-N-oxide
Formula: C₉H₇NO
Boiling Pt: 172 °C
MDL Number: MFCD00006737
CAS Number: 1613-37-2
EINECS: 216-560-6

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR