Ramipril ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydro-cyclopenta[b]pyrrole-2-carboxylic acid, Altace, HOE498, (2S,3aS,6aS)-1-((S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoyl)octahydrocyclopenta[b]pyrrole-2-carboxylic acid

R0219-1G R0219-5G
TCR0219-1GEA 111 USD
TCR0219-1G TCR0219-5G
Ramipril ≥98.0% (by HPLC, titration analysis)
Ramipril
Formula: C₂₃H₃₂N2₂O₅
MW: 416.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00865775
CAS Number: 87333-19-5
Merck Index: 14,08103

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C
Specific rotation [a]20/D +38.0 to +42.0 deg (C=1, 0.1N HCl in EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR