MHY1485 ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,6-Dimorpholino-N-(4-nitrophenyl)-1,3,5-triazin-2-amine

M3107-100MG M3107-25MG
TCM3107-100MGEA 502 USD
TCM3107-100MG TCM3107-25MG
MHY1485 ≥95.0% (by HPLC)
MHY1485
Formula: C₁₇H₂₁N₇O₄
MW: 387.39 g/mol
Melting Pt: 259 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00489974
CAS Number: 326914-06-1

Order Now

Specification Test Results

Appearance White to Amber powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 24.00 to 26.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR