Fulvestrant ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: ICI182,780, (7R,8R,9S,13S,14S,17S)-13-methyl-7-(9-((4,4,5,5,5-pentafluoropentyl)sulphinyl)nonyl)-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol

F1144-100MG F1144-25MG
TCF1144-100MGEA 372 USD
TCF1144-100MG TCF1144-25MG
Fulvestrant ≥98.0% (by HPLC)
Fulvestrant
Formula: C₂₃H₄₇F₅O₃S
MW: 606.77 g/mol
MDL Number: MFCD00903953
CAS Number: 129453-61-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]25 +108 to +115 deg (365nm, C=2, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR