FR180204 ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
FR180204
, FR 180204
, FR-180204
, 5-(2-phenylpyrazolo[1,5-a]pyridin-3-yl)-2H-pyrazolo[3,4-c]pyridazin-3-amine
, 5-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-amine
F1214-5MG
F1214-25MG
TCF1214-25MGEA
420
USD
TCF1214-25MG
TCF1214-5MG
FR180204 ≥98.0% (by HPLC)
FR180204
Formula:
C₁₈H₁₃N₇ MW: 327.34 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14585805 CAS Number: 865362-74-9 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 28.50 to 31.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...