Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer

Supplier: TCI America

H1562-1G H1562-200MG
TCH1562-1GEA 1415 USD
TCH1562-1G TCH1562-200MG
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer
Formula: C₁₆H₂₆O₂Rh₂
MDL Number: MFCD04974244
CAS Number: 73468-85-6

Order Now

Specification Test Results

Elemental analysis (Carbon) 40.00 to 44.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR