Diacetato{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II)

Supplier: TCI America

R0170-1G R0170-200MG
TCR0170-1GEA 194 USD
TCR0170-1G TCR0170-200MG
Diacetato{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II)
Diacetato{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II)
Storage Temperature: Refrigerator
MDL Number: MFCD09753022
CAS Number: 374067-50-2

Order Now

Specification Test Results

IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR