2-Diphenylphosphino-6-methylpyridine ≥95.0% (by GC)

Supplier: TCI America
Warning

D5683-1G D5683-5G
TCD5683-1GEA 114 USD
TCD5683-1G TCD5683-5G
2-Diphenylphosphino-6-methylpyridine ≥95.0% (by GC)
2-Diphenylphosphino-6-methylpyridine
Formula: C₁₈H₁₆NP
MDL Number: MFCD04974234
CAS Number: 132682-77-0

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 93.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR