DBeQ ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N,N'-Dibenzylquinazoline-2,4-diamine, N2,N4-Dibenzylquinazoline-2,4-diamine

D5476-100MG D5476-25MG
TCD5476-100MGEA 278 USD
TCD5476-100MG TCD5476-25MG
DBeQ ≥98.0% (by HPLC)
DBeQ
Formula: C₂₂H₂₀N₄
MW: 340.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03691820
CAS Number: 177355-84-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.50 to 17.00 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR