KN-93 ≥98.0% (by HPLC)

Supplier: TCI America

K0055-25MG K0055-5MG
TCK0055-25MGEA 420 USD
TCK0055-25MG TCK0055-5MG
KN-93 ≥98.0% (by HPLC)
KN-93
Formula: C₂₆H₂₉ClN₂O₄S
MW: 501.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236424
CAS Number: 139298-40-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.00 to 6.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR