Methylpropyldichlorosilane ≥97.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Silane, dichloromethylpropyl-, dichloro(methyl)(propyl)silane, Dichloro(methyl)propylsilane

D5568-25G D5568-5G
TCD5568-25GEA 1156 USD
TCD5568-25G TCD5568-5G
Methylpropyldichlorosilane ≥97.0% (by GC)
Methylpropyldichlorosilane
Formula: C₄H₁₀Cl₂Si
MW: 157.109 g/mol
Boiling Pt: 124 °C
Flash Pt: 27 °C
MDL Number: MFCD00013608
CAS Number: 4518-94-9
EINECS: 224-843-0
UN: 2986
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR