Ethyl azidoacetate ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (2-ethoxy-2-oxoethyl)imino-iminoammonium, Ethyl 2-(1H-triazirin-1-yl)acetate, Acetic acid, 2-azido-, ethyl ester

E1255-25G E1255-5G
TCE1255-25GEA 157 USD
TCE1255-25G TCE1255-5G
Ethyl azidoacetate ≥95.0% (by GC)
Ethyl azidoacetate
Formula: C₄H₇N₃O₂
MW: 129.12 g/mol
Boiling Pt: 164 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190177
CAS Number: 637-81-0
EINECS: 211-301-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR