4-Hydroxy-7-methylcoumarin ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
H1570-5G
H1570-1G
TCH1570-5GEA
92
USD
TCH1570-5G
TCH1570-1G
4-Hydroxy-7-methylcoumarin ≥98.0% (by GC)
4-Hydroxy-7-methylcoumarin
Formula:
C₁₀H₈O₃ MW: 176.17 g/mol |
MDL Number:
MFCD03412316 CAS Number: 18692-77-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...