Sildenafil citrate ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: 5-[2-Ethoxy-5-[(4-methylpiperazin-1-yl)sulphonyl]phenyl]-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one Citrate, 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulphonyl]-4-methylpiperazine Citrate

S0986-100MG S0986-1G S0986-25MG
TCS0986-100MGEA 42 USD
TCS0986-100MG TCS0986-1G TCS0986-25MG
Sildenafil citrate ≥98.0% (by HPLC)
Sildenafil citrate
Formula: C₂₂H₃₀N₆O₄S·C₆H₈O₇
MW: 666.7 g/mol
MDL Number: MFCD09026931
CAS Number: 171599-83-0
Merck Index: 14,08489

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR