Tris(trimethylsiloxy)silane ≥98.0% (by GC)

Supplier: TCI America
Warning

T3520-25ML T3520-5ML
TCT3520-25MLEA 156 USD
TCT3520-25ML TCT3520-5ML
Tris(trimethylsiloxy)silane ≥98.0% (by GC)
Tris(trimethylsiloxy)silane
Formula: C₉H₂₈O₃Si₄
Boiling Pt: 185 °C
Flash Pt: 55 °C
MDL Number: MFCD00054865
CAS Number: 1873-89-8
EINECS: 217-497-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR