Titanium(IV) 2-ethylhexyloxide

Supplier: TCI America
Warning

T3682-100G T3682-500G
TCT3682-100GEA 25 USD
TCT3682-100G TCT3682-500G
Titanium(IV) 2-ethylhexyloxide
Titanium(IV) 2-ethylhexyloxide
Formula: C₃₂H₆₈O₄Ti
Flash Pt: 53 °C
MDL Number: MFCD00015264
CAS Number: 1070-10-6
EINECS: 213-969-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Ignition residue 13.4 to 14.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR