Propargyl ether ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Dipropargylether

P2335-1G P2335-5G
TCP2335-1GEA 75 USD
TCP2335-1G TCP2335-5G
Propargyl ether ≥98.0% (by GC)
Propargyl ether
Formula: C₆H₆O
MW: 94.11 g/mol
Boiling Pt: 117 °C
Flash Pt: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048108
CAS Number: 6921-27-3
UN: 3271
ADR: 3 III

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR