Polyethylene glycol methyl ether methacrylate n=∼9 stabilised
Supplier: TCI America
Synonyms:
Polyethylene glycol monomethyl ether Methacrylate, mPEG Methacrylate, Methoxy PEG Methacrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
P2699-25G
P2699-500G
TCP2699-25GEA
37
USD
TCP2699-25G
TCP2699-500G
Polyethylene glycol methyl ether methacrylate n=∼9 stabilised
Polyethylene glycol methyl ether methacrylate
Formula:
CH₃(OCH₂CH₂)nOCOC(=CH₂)CH₃ |
MDL Number:
MFCD00241432 CAS Number: 26915-72-0 |
Specification Test Results
Appearance | Colourless to Light yellow clear liquid |
The average value of n | 8.5 to 9.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...