Polyethylene glycol methyl ether acrylate n=∼9 stabilised

Supplier: TCI America

Synonyms: mPEG-acrylate, Polyethylene glycol monomethyl ether acrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2698-25G P2698-500G
TCP2698-25GEA 39.1 USD
TCP2698-25G TCP2698-500G
Polyethylene glycol methyl ether acrylate n=∼9 stabilised
Polyethylene glycol methyl ether acrylate
Formula: CH₃(OCH₂CH₂)nOCOC=CH₂
MDL Number: MFCD00803685
CAS Number: 32171-39-4

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
The average value of n 8.5 to 9.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR