Polyethylene glycol diacrylate (PEGDA) n=∼9 stabilized
Supplier: TCI America
|
Danger
|
Synonyms:
PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
P2708-25G
P2708-500G
TCP2708-25GEA
25
USD
TCP2708-25G
TCP2708-500G
Polyethylene glycol diacrylate (PEGDA) n=∼9 stabilized
Polyethylene glycol diacrylate (PEGDA)
Formula:
CH₂=COC(OCH₂CH₂)nOCOC=CH₂ Flash Pt: 104 °C |
MDL Number:
MFCD00081876 CAS Number: 26570-48-9 |
Specification Test Results
Appearance | Colourless to Light yellow clear liquid |
The average value of n | 8.5 to 9.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...