Fluorene-2-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 9H-Fluorene-2-boronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

F1245-25G F1245-5G
TCF1245-25GEA 203 USD
TCF1245-25G TCF1245-5G
Fluorene-2-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Fluorene-2-boronic acid pinacol ester
Formula: C₁₉H₂₁BO₂
MW: 292.18 g/mol
MDL Number: MFCD16294548
CAS Number: 922706-40-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR