Ethyl acetoacetate propylene glycol ketal ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl Acetoacetate Propylene glycol Ketal, 2-(2,4-Dimethyl-1,3-dioxolan-2-yl)acetic acid ethyl ester, Ethyl 2-(2,4-Dimethyl-1,3-dioxolan-2-yl)acetate (mixture of diastereoisomers)

E1417-100G E1417-25G
TCE1417-100GEA 80 USD
TCE1417-100G TCE1417-25G
Ethyl acetoacetate propylene glycol ketal ≥98.0% (by GC)
Ethyl acetoacetate propylene glycol ketal
Formula: C₉H₁₆O₄
MW: 188.23 g/mol
Boiling Pt: 76 °C
Flash Pt: 65 °C
MDL Number: MFCD00151819
CAS Number: 6290-17-1
EINECS: 228-536-2

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 93.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR