Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III) ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: [(ppy)2IrCl]2, Dichlorotetrakis[2-(2-pyridinyl)phenyl]diiridium(III), Dichlorotetrakis(2-(2-pyridinyl)phenyl)diiridium(III)

D5744-1G D5744-200MG
TCD5744-1GEA 449 USD
TCD5744-1G TCD5744-200MG
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III) ≥98.0% (by HPLC)
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
Formula: C₄₄H₃₂Cl₂Ir₂N₄
MW: 1072.09 g/mol
MDL Number: MFCD07784515
CAS Number: 92220-65-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR