1,4-Benzenediacrylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1,4-Phenylenediacrylic acid, 1,4-Benzenediacrylic acid
P1309-1G
P1309-5G
TCP1309-1GEA
17
USD
TCP1309-1G
TCP1309-5G
1,4-Benzenediacrylic acid ≥98.0% (by GC, titration analysis)
1,4-Benzenediacrylic acid
Formula:
C₁₂H₁₀O₄ MW: 218.21 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD00002698 CAS Number: 16323-43-6 EINECS: 240-399-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...